CAS RN 439288-66-1
IUPAC Nombre:
N-(benzenesulfonyl)-2-(4-{4,9-diethoxy-1-oxo-1H,2H,3H-benzo[f]isoindol-2-yl}phenyl)acetamide
Sinónimos:
N-(benzenesulfonyl)-2-(4-{4,9-diethoxy-1-oxo-3H-benzo[f]isoindol-2-yl}phenyl)acetamide
Peso molecular (g/mol):
544.62
Fórmula molecular:
C30H28N2O6S
InChi Key:
XREWXJVMYAXCJV-UHFFFAOYSA-N
SMILES:
CCOC1=C2C=CC=CC2=C(OCC)C2=C1CN(C2=O)C1=CC=C(CC(=O)NS(=O)(=O)C2=CC=CC=C2)C=C1
1
–
1
de
1
resultados
1
GW627368, MedChemExpress
MedChemExpress GW627368 (GW627368X) is a novel, potent and selective competitive antagonist of prostanoid EP4 receptor with additional human TP receptor affinity, with pKi values of 7.0 and 6.8 for human prostanoid EP4 and TP receptors respectively.