CAS RN 425399-05-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CASIN, MedChemExpress
MedChemExpress CASIN is a selective GTPase Cdc42 inhibitor with an IC50 of 2 uM. CASIN can be used for the research of cancer.