CAS RN 4237-93-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress ACTH (11-24)
MedChemExpress ACTH (11-24) is a fragment of adrenocorticotrophin, acts as an antagonist of adrenocorticotropic hormone (ACTH) receptor, and induces cortisol release.
2
MedChemExpress ACTH (11-24)
MedChemExpress ACTH (11-24) is a fragment of adrenocorticotrophin, acts as an antagonist of adrenocorticotropic hormone (ACTH) receptor, and induces cortisol release.
3
MedChemExpress ACTH (11-24)
MedChemExpress ACTH (11-24) is a fragment of adrenocorticotrophin, acts as an antagonist of adrenocorticotropic hormone (ACTH) receptor, and induces cortisol release.