CAS RN 420126-30-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Antitumor agent-3, MedChemExpress
MedChemExpress Antitumor agent-3 is a potent compound which comprises a new imidazopyridine having excellent antitumor activity as an active ingredient.