CAS RN 41964-07-2
IUPAC Nombre:
5-(3-methylphenoxy)-1,2-dihydropyrimidin-2-one
Sinónimos:
tolimidone
Peso molecular (g/mol):
202.21
Fórmula molecular:
C11H10N2O2
InChi Key:
HJQILFPVRNHTIG-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(OC2=CNC(=O)N=C2)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ MLR 1023
CAS: 41964-07-2 Fórmula molecular: C11H10N2O2
2
Tolimidone, MedChemExpress
MedChemExpress Tolimidone is a potent and selective allosteric activator of Lyn kinase with an EC50 of 63 nM.