CAS RN 41552-82-3
IUPAC Nombre:
(3R,4S,5R)-2-[6-(cyclopentylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Sinónimos:
N6-cyclopentyladenosine
Peso molecular (g/mol):
335.36
Fórmula molecular:
C15H21N5O4
InChi Key:
SQMWSBKSHWARHU-FJFSNTMWSA-N
SMILES:
OC[C@H]1OC([C@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCC3)N=CN=C12
1
–
2
de
2
resultados
2
N6-Cyclopentyladenosine, MedChemExpress
MedChemExpress N6-Cyclopentyladenosine (CPA) is a selective Adenosine A1 receptor agonist, with Ki values of 2.3 nM, 790 nM and 43 nM for human A1, A2A and A3 receptors, respectively.