CAS RN 41276-02-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TIC10 isomer, MedChemExpress
MedChemExpress TIC10 isomer is the isomer of TIC10. TIC10 isomer does not possess the reported biological activity of inducing TRIAL expression.