CAS RN 40567-80-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TOPS, MedChemExpress
MedChemExpress TOPS, a Trinder's reagent, is a novel highly water-soluble aniline derivative; are widely used in diagnostic tests and biochemical tests.