CAS RN 3966-30-1
IUPAC Nombre:
2-hydroxy-2-phenylpropanoic acid
Sinónimos:
(+-)-atrolactic acid (1)-2-phenyllactic acid hydroxymethylmandelic acid atrolactic acid, (+-)- atrolactinic acid dl-atrolactic acid (+-)-isomer atrolactic acid dl-atrolactate (S)-atrolactic acid atrolactinate
Peso molecular (g/mol):
166.18
Fórmula molecular:
C9H10O3
InChi Key:
NWCHELUCVWSRRS-UHFFFAOYNA-N
SMILES:
CC(O)(C(O)=O)C1=CC=CC=C1
1
–
1
de
1
resultados
1
Ácido (R)-(-)-2-hidroxi-2-fenilpropiónico, + 98 %, Thermo Scientific Chemicals
CAS: 3966-30-1 Fórmula molecular: C9H10O3 Peso molecular (g/mol): 166.176 Número MDL: MFCD00067699 Clave InChI: NWCHELUCVWSRRS-SECBINFHSA-N Sinónimo: r---2-hydroxy-2-phenylpropionic acid,2r-2-hydroxy-2-phenylpropanoic acid,unii-h38dkr9931,r-2-hydroxy-2-phenylpropanoic acid,phenyllactic acid,atrolactic acid,-,2-hydroxy-2-phenylpropanoic acid #,r---atrolactic acid,r-phenyl lactic acid,r-alpha-hydroxy-alpha-methylbenzeneacetic acid PubChem CID: 736858 Nombre IUPAC: ácido (2R)-2-hidroxi-2-fenilpropanoico SMILES: CC(C1=CC=CC=C1)(C(=O)O)O