CAS RN 39122-38-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC 185058, MedChemExpress
MedChemExpress NSC 185058 is an inhibitor of ATG4B, a major cysteine protease. Inhibition of ATG4B using NSC 185058 markedly attenuates autophagic activity.