CAS RN 38234-21-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Fertirelin
MedChemExpress Fertirelin is a GnRH and LH-RH analogue; it also becomes the treatment choice for reversing cow follicular cysts.
2
MedChemExpress Fertirelin
MedChemExpress Fertirelin is a GnRH and LH-RH analogue; it also becomes the treatment choice for reversing cow follicular cysts.
3
MedChemExpress Fertirelin
MedChemExpress Fertirelin is a GnRH and LH-RH analogue; it also becomes the treatment choice for reversing cow follicular cysts.
4
MedChemExpress Fertirelin
MedChemExpress Fertirelin is a GnRH and LH-RH analogue; it also becomes the treatment choice for reversing cow follicular cysts.