CAS RN 37860-51-8
IUPAC Nombre:
2-[2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethoxy]ethyl 4-methylbenzene-1-sulfonate
Sinónimos:
2-[2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethoxy]ethyl 4-methylbenzenesulfonate
Peso molecular (g/mol):
502.59
Fórmula molecular:
C22H30O9S2
InChi Key:
SLAONPBUWDUSSO-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
1
–
1
de
1
resultados
1
MedChemExpress Bis-Tos-PEG4
MedChemExpress Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.