CAS RN 37661-08-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bacampicillin hydrochloride, MedChemExpress
MedChemExpress Bacampicillin hydrochloride is a penicillin antibiotic, is a prodrug of ampicillin with improved oral bioavailability.