CAS RN 36062-07-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Octahydrocurcumin, MedChemExpress
MedChemExpress Octahydrocurcumin is a hydrogenated derivatives of curcumin; metabolite of curcumin.