CAS RN 356559-20-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SB 525334, MedChemExpress
MedChemExpress SB 525334 is a potent and selective transforming growth factor β1 receptor (ALK5) inhibitor with an IC50 of 14.3 nM.