CAS RN 3546-41-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pyrvinium pamoate, MedChemExpress
MedChemExpress Pyrvinium pamoate is an FDA-approved antihelmintic drug that inhibits WNT pathway signaling.