CAS RN 350997-39-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ O8 OGG1 Inhibitor
Potent and selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 0.2 μM in a gel based assay)