CAS RN 350-12-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sulbentine, MedChemExpress
MedChemExpress Sulbentine (Dibenzthione) is an azole antifungal agent that has fungistatic and fungicidal activities. Sulbentine is used as a locally acting antimycotic in vivo.