CAS RN 349438-38-6
IUPAC Nombre:
N-[(2H-1,3-benzodioxol-5-yl)methyl]-2,6-dichlorobenzamide
Sinónimos:
N-(2H-1,3-benzodioxol-5-ylmethyl)-2,6-dichlorobenzamide
Peso molecular (g/mol):
324.16
Fórmula molecular:
C15H11Cl2NO3
InChi Key:
NMKJFZCBCIUYHI-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(Cl)=C1C(=O)NCC1=CC2=C(OCO2)C=C1
1
–
2
de
2
resultados
1
Alda-1, MedChemExpress
MedChemExpress Alda-1 is a potent and selective ALDH2 agonist, which activates wild-type ALDH2 and restores near wild-type activity to ALDH2*2.
2
Tocris Bioscience™ Alda 1
CAS: 349438-38-6 Fórmula molecular: C15H11Cl2NO3