CAS RN 342639-96-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
KRIBB11, MedChemExpress
MedChemExpress KRIBB11 is an inhibitor of Heat shock factor 1 (HSF1), with IC50 of 1.2 μM.
2
Tocris Bioscience™ KRIBB11
HSF1 inhibitor
3
4