CAS RN 34023-62-6
IUPAC Nombre:
1-(2H-1,3-benzodioxole-5-carbonyl)piperidine
Sinónimos:
1-(2H-1,3-benzodioxole-5-carbonyl)piperidine
Peso molecular (g/mol):
233.27
Fórmula molecular:
C13H15NO3
InChi Key:
BXBNADAPIHHXJQ-UHFFFAOYSA-N
SMILES:
O=C(N1CCCCC1)C1=CC=C2OCOC2=C1
1
–
1
de
1
resultados
1
1-BCP, MedChemExpress
MedChemExpress 1-BCP (Piperonylic acid piperidide) is a centrally active drug that modulates AMPA receptor gated currents. 1-BCP is a memory-enhancing agent.