CAS RN 334951-90-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BIBF0775, MedChemExpress
MedChemExpress BIBF0775 is a potent and selective transforming growth factor β (TGFβ) type I receptor (Alk5) inhibitor with an IC50 of 34 nM.