CAS RN 334-49-6
IUPAC Nombre:
(2E)-dec-2-enoic acid
Sinónimos:
trans-2-decenoic acid 2-decylenic acid (E)-2-decensaeure
Peso molecular (g/mol):
170.25
Fórmula molecular:
C10H18O2
InChi Key:
WXBXVVIUZANZAU-CMDGGOBGSA-N
SMILES:
CCCCCCC\C=C\C(O)=O
1
–
1
de
1
resultados
1
(E)-2-Decenoic acid, MedChemExpress
MedChemExpress (E)-2-Decenoic acid is an interesting fatty acid isolated from royal jelly secretions of honey bees.