CAS RN 333745-53-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LX2343, MedChemExpress
MedChemExpress LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance.