CAS RN 329773-35-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Cinaciguat, MedChemExpress
MedChemExpress Cinaciguat is an activator of guanylate cyclase (sGC), and used for acute decompensated heart failure.