CAS RN 329221-38-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AX20017, MedChemExpress
MedChemExpress AX20017 is a small-molecule protein kinase G (PknG) inhibitor with an IC50 of 0.39 μM.