CAS RN 317366-82-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress β-Amyloid (42-1), human
MedChemExpress β-Amyloid (42-1), human is the inactive form of Amyloid β Peptide (1-42). β-Amyloid (42-1), human is a 42-amino acid peptide which plays a key role in the pathogenesis of Alzheimer disease.