CAS RN 316791-23-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress Thymus peptide C
MedChemExpress Thymus peptide C is a hormonal drug derived from the thymus glands of young calves, which works as a substitute for the physiological functions of the thymus.