CAS RN 31645-39-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Palifosfamide, MedChemExpress
MedChemExpress Palifosfamide is a novel DNA alkylator and the active metabolite of ifosfamide, with antitumor activity.