CAS RN 314045-39-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ BPTES
Glutaminase Inhibitors
2
BPTES, MedChemExpress
MedChemExpress BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM.