CAS RN 312951-85-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PROTO-1, MedChemExpress
MedChemExpress PROTO-1 shows significant protection of hair cells, with HC50(concentration that would produce 50% hair cell survival) of 1 (mu)M-10 (mu)M (1 (mu)M≤HC50≤10 (mu)M).