CAS RN 312917-14-9
IUPAC Nombre:
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
Sinónimos:
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
Peso molecular (g/mol):
332.42
Fórmula molecular:
C20H16N2OS
InChi Key:
WQGDQGAFSDMBLA-UHFFFAOYSA-N
SMILES:
O=C(NC1=C(C#N)C2=C(CCCC2)S1)C1=C2C=CC=CC2=CC=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ TCS JNK 5a
CAS: 312917-14-9 Fórmula molecular: C20H16N2OS
2
TCS JNK 5a, MedChemExpress
MedChemExpress TCS JNK 5a is a potent JNK3 inhibitor with a pIC50 of 6.7. TCS JNK 5a also inhibits JNK2 with a pIC50 of 6.5.