CAS RN 312756-74-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
RS-1, MedChemExpress
MedChemExpress RS-1 is a RAD51 activator, and also increases CRISPR/Cas9-mediated knock-in efficiencies.
2