CAS RN 307-60-8
IUPAC Nombre:
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-12-iodododecane
Sinónimos:
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-12-iodododecane
Peso molecular (g/mol):
746
Fórmula molecular:
C12F25I
InChi Key:
WBYCUETVRCXUPE-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
1
–
1
de
1
resultados
1
Perfluorododecilo Yoduro, TRC
CAS: 307-60-8 Fórmula molecular: C12 F25 I Peso molecular (g/mol): 745.993 Sinónimo: 1-Iodoperfluorododecane,1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-12-iodododecane,Pentacosafluoro-1-iodododecane Nombre IUPAC: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-12-iodododecano SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I