CAS RN 306288-04-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AVE 0991 sodium salt, MedChemExpress
MedChemExpress AVE 0991 sodium salt is a nonpeptide and orally active Ang-(1-7) receptor Mas agonist. AVE 0991 competes for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 of 21 nM.