CAS RN 303149-14-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HT-2157, MedChemExpress
MedChemExpress HT-2157 (SNAP 37889) is a selective, high-affinity, competitive antagonists of galanin-3 receptor (Gal3).