CAS RN 301836-41-9
IUPAC Nombre:
4-[4-(2H-1,3-benzodioxol-5-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Sinónimos:
4-[4-(2H-1,3-benzodioxol-5-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Peso molecular (g/mol):
384.4
Fórmula molecular:
C22H16N4O3
InChi Key:
FHYUGAJXYORMHI-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=N1)C1=CC2=C(OCO2)C=C1
1
–
4
de
4
resultados
1
Promociones disponibles
Tocris Bioscience™ SB 431542
Tocris Bioscience™ SB 431542
Potent, selective inhibitor of TGF-βRI, ALK4 and ALK7
2
SB 431542, TRC
CAS: 301836-41-9 Fórmula molecular: C22 H16 N4 O3 Peso molecular (g/mol): 384.39 Sinónimo: SB 431542,4-[4-(1,3-Benzodioxol-5-yl)-5-(2-pyridinyl)-1H-imidazol-2-yl]-benzamide Nombre IUPAC: 4-[4-(1,3-benzodioxol-5-il)-5-piridina-2-il-1H-imidazol-2-il]benzamida SMILES: NC(=O)c1ccc(cc1)c2nc(c3ccc4OCOc4c3)c([nH]2)c5ccccn5
3
SB-431542, MedChemExpress
MedChemExpress SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM.
4
STEMCELL Technologies SB431542 (Hydrate)
Activin/BMP/TGFβ pathway inhibitor; Inhibits ALK4, ALK5, and ALK7