CAS RN 298684-44-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
ML402, MedChemExpress
MedChemExpress ML402, a thiophene-carboxamide, is a selective K2P2.1(TREK-1) and K2P10.1(TREK-2) activator. ML402 is inactive against K2P4.1(TRAAK).
2
Tocris™ ML 402
Selective TREK channel activator