CAS RN 293753-05-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SR3335, MedChemExpress
MedChemExpress SR3335 (ML 176) is a selective RORα inverse agonist that directly binds to RORα with a Ki of 220 nM.