CAS RN 285986-88-1
IUPAC Nombre:
2-({[3,5-bis(trifluoromethyl)phenyl]formamido}oxy)-N-(4-chlorophenyl)propanamide
Sinónimos:
2-({[3,5-bis(trifluoromethyl)phenyl]formamido}oxy)-N-(4-chlorophenyl)propanamide
Peso molecular (g/mol):
454.75
Fórmula molecular:
C18H13ClF6N2O3
InChi Key:
DSMXVSGJIDFLKP-UHFFFAOYNA-N
SMILES:
CC(ONC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(=O)NC1=CC=C(Cl)C=C1
1
–
1
de
1
resultados
1
CCG-1423, MedChemExpress
MedChemExpress CCG-1423 is a novel inhibitor of RhoA/C-mediated gene transcription that is capable of inhibiting invasion of PC-3 prostate cancer cells in a Matrigel model of metastasis.