CAS RN 2856-75-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Modaline sulfate, MedChemExpress
MedChemExpress Modaline sulfate is a MAO inhibitor, used in the treatment of depression.