CAS RN 284490-13-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Clorhidrato de fodosina, TRC
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
2
Forodesine hydrochloride, MedChemExpress
MedChemExpress Forodesine hydrochloride (BCX-1777 hydrochloride) is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor with IC50 values ranging from 0.48 to 1.57 nM for human, mouse, rat, monkey and dog PNP. Forodesine hydrochloride is a potent human lymphocyte proliferation inhibitor. Forodesine hydrochloride could induce apoptosis in leukemic cells by increasing the dGTP levels.