CAS RN 280744-09-4
IUPAC Nombre:
3-(2,4-dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Sinónimos:
3-(2,4-dichlorophenyl)-4-(1-methylindol-3-yl)-1H-pyrrole-2,5-dione
Peso molecular (g/mol):
371.22
Fórmula molecular:
C19H12Cl2N2O2
InChi Key:
JCSGFHVFHSKIJH-UHFFFAOYSA-N
SMILES:
CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CC=C(Cl)C=C1Cl
1
–
3
de
3
resultados
1
Tocris Bioscience™ SB 216763
CAS: 280744-09-4 Fórmula molecular: C19H12Cl2N2O2
2
3
SB 216763, MedChemExpress
MedChemExpress SB 216763 is potent, selective and ATP-competitive GSK-3 inhibitor with IC50s of 34.3 nM for both GSK-3α and GSK-3β.