CAS RN 27599-63-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress 5(6)-Aminofluorescein
MedChemExpress 5(6)-Aminofluorescein (5(6)-AFM) is a a precursor for synthesis of 5(6)-FITC (HY-15941). 5(6)-FITC is an amine-reactive derivative of fluorescein dye.