CAS RN 2719-05-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FH1, MedChemExpress
MedChemExpress FH1 (NSC 12407) enhances hepatocyte functions, and promotes the differentiation of induced pluripotent stem (iPS)-derived hepatocytes toward a phenotype more mature and the maturation of well-differentiated cultures of hepatocyte-like cells (iHeps).