CAS RN 2634129-34-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Janelia Fluor™ 552, Haloalkane
Cell-permeable fluorogenic dye with a chloroalkane handle. Application: live-cell imaging as a self-labeling tag substrate. Also chemigenetic Ca2+ indicator.