CAS RN 263251-78-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tavilermide, MedChemExpress
MedChemExpress Tavilermide is a selective, partial agonist of TrkA, or a nerve growth factor (NGF) mimetic.