CAS RN 261365-11-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ MB 05032
Potent FBPase inhibitor; promotes HSC expansion
2
MB05032, MedChemExpress
MedChemExpress MB05032 is a special and efficacious gluconeogenesis inhibitor targeted the AMP binding site of fructose 1,6-bisphosphatase (FBPase) with an IC50 value of 16 nM.