CAS RN 2608-21-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sotetsuflavone, MedChemExpress
MedChemExpress Sotetsuflavone is a potent inhibitor of DENV-NS5 RdRp (Dengue virus NS5 RNA-dependent RNA polymerase) with an IC50 of 0.16 uM, is the most active compound of this series .