CAS RN 260430-02-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress Cy2
MedChemExpress CY2 Non-Sulfonated (Cyanine2) is a dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides.